
Atorvastatin Related Compound A

Synonym(s):
(βR,δR)-β,δ-Dihydroxy-2-(1-methylethyl)-4,5-diphenyl-3-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic acid hemicalcium salt
Empirical Formula (Hill Notation):
C66H70CaN4O10
CAS Number:
Molecular Weight:
1119.36
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2