
Articaine Related Compound A

Synonym(s):
Methyl 4-methyl-3-[2-(propylamino)acetamido]-2-thiophenecarboxylate, Methyl 4-methyl-3-[[2-(propylamino)acetyl]amino]-thiophene-2-carboxylate
Empirical Formula (Hill Notation):
C12H18N2O3S
CAS Number:
Molecular Weight:
270.35
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2