Skip to Content
CX
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Arachidic acid
Synonym(s):
Arachic acid, Arachidinic acid, Eicosanoic acid, Icosanoic acid, NSC 93983
Linear Formula:
CH
3
(CH
2
)
18
COOH
CAS Number:
506-30-9
Molecular Weight:
312.53
EC Number:
208-031-3
Products
Product Category
lipids (2)
building blocks (1)
fatty acids (1)
metabolites (1)
Brand
Sigma-Aldrich (2)
Supelco (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Puriss (1)
analytical standard (1)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
A3631
Arachidic acid,
≥99%
Expand
View Pricing
10930
Arachidic acid,
synthetic, ≥99.0% (GC)
Expand
View Pricing
PHR3530
Arachidic acid,
certified reference material, pharmaceutical secondary standard
Expand
View Pricing
39383
Arachidic acid,
analytical standard
Expand
View Pricing