
Anti-ITIH4 antibody produced in rabbit

Synonym(s):
Anti-GP120, Anti-IHRP, Anti-ITI heavy chain H4, Anti-Inter-alpha-inhibitor heavy chain 4, Anti-Inter-alpha-trypsin inhibitor family heavy chain-related protein, Anti-MUSTN1 antibody produced in rabbit, Anti-PK-120, Anti-Plasma kallikrein sensitive glycoprotein 120, Anti-Transmembrane protein 110 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing