
Ancymidol

Synonym(s):
α-Cyclopropyl-α-(4-methoxyphenyl)-5-pyrimidinemethanol
Empirical Formula (Hill Notation):
C15H16N2O2
CAS Number:
Molecular Weight:
256.30
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
Special Grade
Technique
Showing 1-2 of 2