Skip to Content
Merck
Amoxapine Pharmaceutical Secondary Standard; Certified Reference Material

Amoxapine

Amoxapine Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2-Chloro-11-(1-piperazinyl)dibenz[b,f][1,4]oxazepine, Amoxapine
Empirical Formula (Hill Notation):
C17H16ClN3O
CAS Number:
Molecular Weight:
313.78
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade