
Amlodipine impurity B

Synonym(s):
3-Ethyl 5-methyl (4RS)-4-(2-chlorophenyl)-6-methyl-2-[[2-[[2- (methylcarbamoyl)benzoyl]amino]ethoxy]methyl]-1,4-dihydropyridine-3,5-dicarboxylate, 3-Ethyl 5-methyl 4-(2-chlorophenyl)-6-methyl-2-({2-[2-(methylcarbamoyl)benzamido]ethoxy}methyl)-1,4-dihydropyridine-3,5-dicarboxylate
Empirical Formula (Hill Notation):
C29H32ClN3O7
CAS Number:
Molecular Weight:
570.03
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2