
α-Allocryptopine

Synonym(s):
Allocryptopine, α-Fagarine, β-Homochelidonine, Thalictrimine
Empirical Formula (Hill Notation):
C21H23NO5
CAS Number:
Molecular Weight:
369.41
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing