
5-Bromo-3-fluoropyridine-2-carbonitrile

Synonym(s):
5-Bromo-3-fluoropicolinonitrile
Empirical Formula (Hill Notation):
C6H2BrFN2
CAS Number:
Molecular Weight:
201.00
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing