
4-Ethynylbenzoic acid

Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing