
2-tert-Butyl-4-hydroxyanisole

Synonym(s):
2-BHA, 3-(1,1-Dimethylethyl)-4-methoxyphenol, 3-tert-Butyl-4-methoxyphenol, 2-tert-Butyl-4-hydroxyanisole
Empirical Formula (Hill Notation):
C11H16O2
CAS Number:
Molecular Weight:
180.24
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing