Skip to Content
Merck
All Photos(1)

Key Documents

SML3712

Sigma-Aldrich

NG-497

≥98% (HPLC)

Synonym(s):

6-(4-Ethoxyphenyl)-4-methoxy-2-pyridinecarboxylic acid, 1-methylethyl ester, Isopropyl 6-(4-ethoxyphenyl)-4-methoxypicolinate, NG 497, NG497

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO4
CAS Number:
Molecular Weight:
315.36
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC(OC)=CC(C2=CC=C(OCC)C=C2)=N1)OC(C)C

Biochem/physiol Actions

NG-497 is a non-cytotoxic, substrate-competitive, reversible and highly selective human/primate adipose triglyceride lipase (ATGL; IPLA2-zeta; PNPLA2) inhibitor (IC50 = 1 μM, Ki = 0.5 μM) with little activity toward related human lipid hydrolases or ATGL orthologues from other species (<20% inhibition of mouse, rat, goat, pig, dog, marmoset ATGL at 50 μM). NG-497 abolishes hormone-induced lipolysis in human adipocytes (IC50 = 1.5 and 0.5 μM against 1 μM isoproterenol-induced total or hormonesensitive lipase (HSL)-independent FA release in Simpson-Golabi-Behmel syndrome adipocytes), but not in HepG2 that lacks ATGL hydrolase activity.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gernot F Grabner et al.
Journal of the American Chemical Society, 144(14), 6237-6250 (2022-04-02)
Chronically elevated circulating fatty acid levels promote lipid accumulation in nonadipose tissues and cause lipotoxicity. Adipose triglyceride lipase (ATGL) critically determines the release of fatty acids from white adipose tissue, and accumulating evidence suggests that inactivation of ATGL has beneficial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service