Skip to Content
Merck
All Photos(1)

Documents

PHG0021

SAFC

Choline chloride

Pharma Manufacturing

Synonym(s):

(2-Hydroxyethyl)trimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(Cl)CH2CH2OH
CAS Number:
Molecular Weight:
139.62
Beilstein:
3563126
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder

mol wt

Mw 139.62 g/mol

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial and lead, tested

color

white

pH

5.0-6.5 (25 °C, 140 g/L)

mp

302-305 °C (dec.) (lit.)

solubility

water: soluble 140 g/L

suitability

suitable for (manufacturing use)

foreign activity

foreign activity (Cytotoxicity; tested)

SMILES string

[Cl-].C[N+](C)(C)CCO

InChI

1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1

InChI key

SGMZJAMFUVOLNK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
To request the documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to documentation for this product requires a confidentiality disclosure agreement.

Substrates

Choline acetyltransferase substrate

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service