The Nalgene® Cryoware markers are not alcohol resistant If used on plastics such as wash bottles, When used on poly-cryoware labels, they had a "moderate" tolerance, but would likely not hold up under repeated exposure to alcohols.
M6163
Nalgene® Cryoware marker set
assorted colors, one each red, green, blue, and black
Synonym(s):
cryomarker, cryopen, freezer marker
About This Item
Recommended Products
packaging
pkg of 4
manufacturer/tradename
Nalgene 6313-0010
technique(s)
cryopreservation: suitable
color
assorted colors, one each red, green, blue, and black
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Is this marker resistant to alcohols?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service