PHR3428
Travoprost Related Compound A
certified reference material, pharmaceutical secondary standard
Synonym(s):
(+)-Fluprostenol solution
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1673012
CofA
current certificate can be downloaded
packaging
pkg of 1.2 mL
application(s)
pharmaceutical
format
mixture (Solution in Water : Acetonitrile (55:45))
SMILES string
FC(F)(F)c1cc(ccc1)OC[C@H](O)\C=C\[C@@H]2[C@H]([C@H](C[C@H]2O)O)C\C=C/CCCC(=O)O
InChI
1S/C23H29F3O6/c24-23(25,26)15-6-5-7-17(12-15)32-14-16(27)10-11-19-18(20(28)13-21(19)29)8-3-1-2-4-9-22(30)31/h1,3,5-7,10-12,16,18-21,27-29H,2,4,8-9,13-14H2,(H,30,31)/b3-1-,11-10+/t16-,18-,19-,20+,21-/m1/s1
InChI key
WWSWYXNVCBLWNZ-QIZQQNKQSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
related product
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service