Skip to Content
Merck
All Photos(1)

Documents

M32704

Sigma-Aldrich

2-Methyl-2-butene

≥99%, purified by redistillation

Synonym(s):

β-Isoamylene, Amylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)2
CAS Number:
Molecular Weight:
70.13
Beilstein:
1361353
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.4 (vs air)

Quality Level

vapor pressure

25.09 psi ( 55 °C)
7.66 psi ( 20 °C)

Assay

≥99%

purified by

redistillation

expl. lim.

8.7 %

refractive index

n20/D 1.385 (lit.)

bp

35-38 °C (lit.)

mp

−134 °C (lit.)

density

0.662 g/mL at 25 °C (lit.)

SMILES string

C\C=C(\C)C

InChI

1S/C5H10/c1-4-5(2)3/h4H,1-3H3

InChI key

BKOOMYPCSUNDGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-2-butene can be used:
  • For the cross metathesis reaction to form compounds with prenyl-type double bonds: methyl 10-methylundec-9-enoate, 2-methylundec-2-ene and 8-prenylcoumarins.
  • As a model of C5-branched alkene fuel to study the high-temperature kinetics of branched alkenes in a combustion environment.
  • As an additive along with pyridine for partial hydrogenation of poly-ynes to form ω-hydroxy polyunsaturated fatty acids.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fishing for the right catalyst for the cross-metathesis reaction of methyl oleate with 2-methyl-2-butene.
Sytniczuk A, et al.
Catalysis Science & Technology, 7(6), 1284-1296 (2017)
Influences of the molecular fuel structure on combustion reactions towards soot precursors in selected alkane and alkene flames.
Ruwe L, et al.
Physical Chemistry Chemical Physics, 20(16), 10780-10795 (2018)
Chemical synthesis and biological evaluation of ?-hydroxy polyunsaturated fatty acids.
Hwang SH, et al.
Bioorganic & Medicinal Chemistry Letters, 27(3), 620-625 (2017)
Prenylcoumarins in One or Two Steps by a Microwave-Promoted Tandem Claisen Rearrangement/Wittig Olefination/Cyclization Sequence.
Schultze C and Schmidt B
The Journal of Organic Chemistry, 83(9), 5210-5224 (2018)
Colm Delaney et al.
Lab on a chip, 17(11), 2013-2021 (2017-05-23)
A novel approach that allows control of flow in microfluidic channels with unsurpassed performance using light is described. Valve structures have been created using photoresponsive hydrogels based on spiropyran-functionalised pNIPAAm hydrogels photopolymerised around pillar structures within the channels. Valve actuation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service