Skip to Content
Merck
All Photos(2)

Key Documents

74208

Supelco

Methyl nonadecanoate

analytical standard

Synonym(s):

Nonadecanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17COOCH3
CAS Number:
Molecular Weight:
312.53
Beilstein:
1788184
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

37-40 °C (lit.)
37-41 °C

format

neat

functional group

ester

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C20H40O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-2/h3-19H2,1-2H3

InChI key

BDXAHSJUDUZLDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Methyl nonadecanoate may be used as an internal standard to study the mechanism of anti-oxidation effect of ginseng extract using gas chromatography coupled with electron spin resonance spectrophotometry. Methyl nonadecanoate may also be used as an internal standard to investigate the separation of molecular species of triacylglycerols using high-performance liquid chromatography in silver ion mode.
Methyl nonadecanoate may be used as an internal standard to study the mechanism of anti-oxidation effect of ginseng extract using gas chromatography coupled with electron spin resonance spectrophotometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irene B Gutiérrez et al.
Ecotoxicology and environmental safety, 173, 293-304 (2019-02-19)
S-metolachlor (SMOC) and terbuthylazine (TBA) are herbicides that commonly appear as active ingredients (a.i.) in the composition of plant protection products. In a previous work, experimental bioassays were performed using those chemicals to find suitable molecular biomarkers to assess its
Separation of molecular species of triacylglycerols by high-performance liquid chromatography with a silver ion column
Christie.W.W
Journal of Chromatography A, 454, 273-284 (1988)
Ginseng extract scavenges hydroxyl radical and protects unsaturated fatty acids from decomposition caused by iron-mediated lipid peroxidation
Zhang D, et al.
Free Radical Biology & Medicine, 20(1), 145-150 (1996)
Hala Aswad et al.
Diabetologia, 57(10), 2155-2164 (2014-07-31)
Exosomes released from cells can transfer both functional proteins and RNAs between cells. In this study we tested the hypothesis that muscle cells might transmit specific signals during lipid-induced insulin resistance through the exosomal route. Exosomes were collected from quadriceps

Protocols

Protocol for GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service