Skip to Content
Merck
All Photos(4)

Key Documents

ABE1029

Sigma-Aldrich

Anti-acetyl Histone H3 Antibody (Lys122)

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/I, Histone H3/j, Histone H3/k, Histone H3/l

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (pLys122)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3, also known as Histone H3.1t (H3/t), H3t, H3/g, and encoded by the gene name HIST3H3/ H3FT, is a core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Therefore, histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. High levels of phosphorylation of Histone H3 are associated with mitosis.

Immunogen

Epitope: Near C-terminal
KLH-conjugated linear peptide corresponding to human acetyl Histone H3 (Lys122) near the C-terminal.

Application

Anti-acetyl Histone H3 (Lys122) is an antibody against acetyl Histone H3 (Lys122) for use in WB, ICC, DB, IP.
Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected acetyl Histone H3 (Lys122) in HeLa and A431cells.
Dot Blot Analysis: A 1:1,000 dilution from a representative lot detected acetyl Histone H3 (Lys122) in Histone peptides with various modifications.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated acetyl Histone H3 (Lys122) in 500 µg of sodium butyrate treated HeLa acid extract.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in sodium butyrate treated HeLa acid extract.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected acetyl Histone H3 (Lys122) in 10 µg of sodium butyrate treated HeLa acid extract.

Target description

~17 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service