Skip to Content
Merck
All Photos(1)

Key Documents

8.01987

Sigma-Aldrich

4-tert-Butylpyrocatechol

for synthesis

Synonym(s):

4-tert-Butylpyrocatechol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-[(CH3)3C]C6H3-1,2-(OH)2
CAS Number:
Molecular Weight:
166.22
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-653-9
NACRES:
NA.22

vapor pressure

<1 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

autoignition temp.

160 °C

potency

815 mg/kg LD50, oral (Rat)
1331 mg/kg LD50, skin (Rat)

kinematic viscosity

100 cSt(57 °C)

bp

284-286 °C/1013 hPa

mp

52-55 °C

transition temp

flash point 130 °C

density

1.05 g/cm3 at 60 °C

bulk density

650‑700 kg/m3

storage temp.

2-30°C

InChI

1S/C10H14O2/c1-10(2,3)7-4-5-8(11)9(12)6-7/h4-6,11-12H,1-3H3

InChI key

XESZUVZBAMCAEJ-UHFFFAOYSA-N

Application

  • Stabilization of Soil with 4-Tert-Butylpyrocatechol: Explores the use of 4-tert-butylpyrocatechol as a soil stabilizer to mitigate water′s adverse effects (JB Hemwall, DT Davidson, 2023).
  • Anthranilamide-protected vinylboronic acid: rational monomer design for improved polymerization/transformation ability: Includes a discussion on the protonation of the C–B bond in organic molecules using 4-tert-butylpyrocatechol (H Suzuki et al., 2022).
  • Selective oxidation of 4-tert-butylphenol by hydrogen peroxide in the presence of titanosilicates: Investigates the catalytic transformations of 4-tert-butylphenol into 4-tert-butylpyrocatechol using different catalysts (RR Talipova et al., 2016).
  • A theoretical and experimental study for screening inhibitors for styrene polymerization: Discusses the use of 4-tert-butylpyrocatechol among other compounds as inhibitors in polymerization processes (A Darvishi et al., 2019).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 52 °C
Melting range (upper value): ≤ 57 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service