Skip to Content
Merck
All Photos(1)

Key Documents

870126P

Avanti

16:0 azidocaproyl PE

Avanti Research - A Croda Brand 870126P, powder

Synonym(s):

16:0 azidocaproyl PE, 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(6-azidohexanoyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 g (870126P-1g)
pkg of 1 × 100 mg (870126P-100mg)
pkg of 1 × 25 mg (870126P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870126P

lipid type

click reagents
headgroup modified lipids

shipped in

dry ice

storage temp.

−20°C

General description

16:0 azidocaproyl PE, also known as 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(6-azidohexanoyl) is a head group modified lipid containing an omega-terminal azide. This click reagent with the terminal azide group can be used in a highly specific linking reaction with alkyne-containing reagents, known as ‘click chemistry′, in the presence of a copper (Cu)-containing catalyst.

Application

16:0 azidocaproyl PE or 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(6-azidohexanoyl) has been used as a fluorophore to visualize cellular boundaries of the plasma membrane.

Biochem/physiol Actions

16:0 azidocaproyl PE aids in functionalization of DNA oligonucleotides on supported lipid bilayers for cell biological or in vitro reconstitution studies.

Packaging

20 mL Clear Glass Screw Cap Vial (870126P-1g)
5 mL Amber Glass Screw Cap Vial (870126P-100mg)
5 mL Amber Glass Screw Cap Vial (870126P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kabir H Biswas et al.
Langmuir : the ACS journal of surfaces and colloids, 34(33), 9781-9788 (2018-07-24)
Deoxyribonucleic acid (DNA) has been used as a material for a variety of applications, including surface functionalization for cell biological or in vitro reconstitution studies. Use of DNA-based surface functionalization eliminates limitations of multiplexing posed by traditionally used methods in
Jennifer R Ferrer et al.
ACS nano, 14(2), 1682-1693 (2020-01-18)
Spherical nucleic acids (SNAs) are a class of nanomaterials with a structure defined by a radial distribution of densely packed, short DNA or RNA sequences around a nanoparticle core. This structure allows SNAs to rapidly enter mammalian cells, protects the
Leonhard Möckl et al.
Developmental cell, 50(1), 57-72 (2019-05-21)
The mammalian glycocalyx is a heavily glycosylated extramembrane compartment found on nearly every cell. Despite its relevance in both health and disease, studies of the glycocalyx remain hampered by a paucity of methods to spatially classify its components. We combine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service