Skip to Content
Merck
All Photos(1)

Key Documents

860776P

Avanti

16:0 SB (DPSB)

new

Avanti Research - A Croda Brand

Synonym(s):

Dipalmitoyl-rac-glycero-3-sulfobetaine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H79NO7S
Molecular Weight:
718.12
UNSPSC Code:
12352211
NACRES:
NA.79

Assay

>99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

functional group

(Sphingolipid)

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(C[N+](C)(C)CCCS([O-])(=O)=O)OC(=O)CCCCCCCCCCCCCCC

Application

Targeted drug delivery

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service