Skip to Content
Merck
All Photos(1)

Key Documents

SLW4506/80D

Pyrex® Screwcaps for Media-lab bottle

blue, thread 80 mm

Synonym(s):

Pyrex, SciLabware 4506/80D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

blue
polypropylene

manufacturer/tradename

4506/80D
SciLabware

thread

80 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • For use with SLW1516, SLW1517, SLW1518, SLW1519, SLW1520, SLW1523 and SLW1524 series bottles
  • Polypropylene plug seal cap ideal for media preparation work
  • Autoclavable up to 140 deg. C

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service