Key Documents
DWK34502-99
KIMBLE® KIMAX® open ended melting capillary
conforms to USP, glass, L (100 mm)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material
glass
translucent borosilicate glass tube
sterility
non-sterile
packaging
case of 20 ea 100 tubes in each vial
manufacturer/tradename
Kimble® 34502-99
L
(100 mm)
tube diam.
(1.5-1.8 mm)
General description
Melting Point Capillary
  • KIMAX melting point capillary tubes are used to determine the point at which a solid sample turns to liquid.
  • Tubes are 100 mm long when supplied with both ends open, so that approximately 90 mm will remain after sealing
  • Tube length insures that the open end will extend well above the bath′s surface to prevent the bath liquid from entering the tube
  • 34502-99 and 34507-99 have a smaller I.D. than 34500-99 and are designed from requirements contained in the United States Pharmacopeia (USP)
  • Packaged with 100 tubes per vial and 20 vials per case
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Features and Benefits
• Manufactured from borosilicate glass 5.1• Tubes are 100 mm long when supplied with both ends open, so that approximately 90 mm will remain after sealing• Tube length insures that the open end will extend well above the bath′s surface to prevent the bath liquid from entering the tube• 34502-99 and 34507-99 have a smaller I.D. than 34500-99 and are designed from requirements contained in the United States Pharmacopeia (USP)• Packaged with 100 tubes per vial and 20 vials per case
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.