Skip to Content
Merck
All Photos(1)

Key Documents

C28707

Sigma-Aldrich

4-Chlorobenzenemethanethiol

98%

Synonym(s):

4-Chlorobenzyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2SH
CAS Number:
Molecular Weight:
158.65
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.5893 (lit.)

mp

19-20 °C (lit.)

density

1.202 g/mL at 25 °C (lit.)

SMILES string

SCc1ccc(Cl)cc1

InChI

1S/C7H7ClS/c8-7-3-1-6(5-9)2-4-7/h1-4,9H,5H2

InChI key

GKQXPTHQTXCXEV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ben J Tickner et al.
Chemical science, 10(20), 5235-5245 (2019-06-14)
We report the formation of a series of novel [Ir(H)2(IMes)(α-13C2-carboxyimine)L] complexes in which the identity of the coligand L is varied. When examined with para-hydrogen, complexes in which L is benzylamine or phenethylamine show significant 1H hydride and 13C2 imine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service