Skip to Content
Merck
All Photos(1)

Key Documents

930210

Sigma-Aldrich

N-(2,5-Dichlorobenzoyl)glycyl-L-leucine

≥95.0%

Synonym(s):

(2S)-2-{2-[(2,5-Dichlorophenyl)formamido]acetamido}-4-methylpentanoic acid, CANDDY_MLN Tag

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18Cl2N2O4
CAS Number:
Molecular Weight:
361.22
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

Assay

≥95.0%

form

powder

storage temp.

2-8°C

SMILES string

ClC1=CC=C(Cl)C(C(NCC(N[C@H](C(O)=O)CC(C)C)=O)=O)=C1

Application

CANDDY (Chemical knockdown with Affinity aNd Degradation DYnamics) employs a bispecific molecule (target interactor + linker + CANDDY_MLN) that degrades protein by direct recruitment to the proteasome. Shown to selectively and dose-dependently degrade KRAS G12D/V and MDM2 proteins and suppress their signaling without non-specific cytotoxic effects.

CANDDY, a protein-slaying approach, occurs without ubiquitination and bypasses the need for an E3 ligase. Has the potential to overcome the current limitations of proteolysis and to expand the application scope of degraders.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service