Skip to Content
Merck
All Photos(4)

Key Documents

774448

Sigma-Aldrich

4-Chlorophenylmagnesium bromide solution

greener alternative

1.0 M in 2-methyltetrahydrofuran

Synonym(s):

(4-Chlorophenyl)magnesium bromide, p-Chlorophenylmagnesium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4MgBr
CAS Number:
Molecular Weight:
215.76
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solution

reaction suitability

reaction type: Grignard Reaction

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

1.0 M in 2-methyltetrahydrofuran

density

1.005 g/mL at 25 °C

greener alternative category

storage temp.

2-8°C

SMILES string

Clc1ccc([Mg]Br)cc1

InChI

1S/C6H4Cl.BrH.Mg/c7-6-4-2-1-3-5-6;;/h2-5H;1H;/q;;+1/p-1

InChI key

CDEMHJCJMMOFMB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product, 1.0 M in 2-methyltetrahydrofuran aligns with Safer Solvents and Auxiliaries, Use of Renewable Feedstocks and Inherently Safer Chemistry for Accident Prevention. Find details here.

Application

Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF)

2-Methyltetrahydrofuran (2-MeTHF): A Biomass-Derived Solvent with Broad Application in Organic Chemistry

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

8.6 °F - closed cup

Flash Point(C)

-13.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service