Skip to Content
Merck
All Photos(2)

Key Documents

475203

Sigma-Aldrich

Cyclopentadienylzirconium(IV) trichloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5Cl3Zr
CAS Number:
Molecular Weight:
262.68
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

core: zirconium
reagent type: catalyst

mp

210 °C (dec.) (lit.)

SMILES string

Cl[Zr](Cl)Cl.[CH]1[CH][CH][CH][CH]1

InChI

1S/C5H5.3ClH.Zr/c1-2-4-5-3-1;;;;/h1-5H;3*1H;/q;;;;+3/p-3

InChI key

SULDCQJIUCVYOB-UHFFFAOYSA-K

Application

Catalyst for:
  • Homo- and copolymerization reactions
  • Catalytic dehydrogenation of cyclooctane
  • Cross-coupling reaction of aryl fluorides with a phenethyl Grignard reagents

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service