Merck
Autocrine activation of P2Y1 receptors couples Ca (2+) influx to Ca (2+) release in human pancreatic beta cells.
Diabetologia (2014-09-12)
Shara Khan, Richard Yan-Do, Eric Duong, Xichen Wu, Austin Bautista, Stephen Cheley, Patrick E MacDonald, Matthias Braun
RESUMEN
There is evidence that ATP acts as an autocrine signal in beta cells but the receptors and pathways involved are incompletely understood. Here we investigate the receptor subtype(s) and mechanism(s) mediating the effects of ATP on human beta cells. We examined the effects of purinergic agonists and antagonists on membrane potential, membrane currents, intracellular Ca(2+) ([Ca(2+)]i) and insulin secretion in human beta cells. Extracellular application of ATP evoked small inward currents (3.4 ± 0.7 pA) accompanied by depolarisation of the membrane potential (by 14.4 ± 2.4 mV) and stimulation of electrical activity at 6 mmol/l glucose. ATP increased [Ca(2+)]i by stimulating Ca(2+) influx and evoking Ca(2+) release via InsP3-receptors in the endoplasmic reticulum (ER). ATP-evoked Ca(2+) release was sufficient to trigger exocytosis in cells voltage-clamped at -70 mV. All effects of ATP were mimicked by the P2Y(1/12/13) agonist ADP and the P2Y1 agonist MRS-2365, whereas the P2X(1/3) agonist α,β-methyleneadenosine-5-triphosphate only had a small effect. The P2Y1 antagonists MRS-2279 and MRS-2500 hyperpolarised glucose-stimulated beta cells and lowered [Ca(2+)]i in the absence of exogenously added ATP and inhibited glucose-induced insulin secretion by 35%. In voltage-clamped cells subjected to action potential-like stimulation, MRS-2279 decreased [Ca(2+)]i and exocytosis without affecting Ca(2+) influx. These data demonstrate that ATP acts as a positive autocrine signal in human beta cells by activating P2Y1 receptors, stimulating electrical activity and coupling Ca(2+) influx to Ca(2+) release from ER stores.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Purinergic Receptor P2Y1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder