Merck
MIF Is Necessary for Late-Stage Melanoma Patient MDSC Immune Suppression and Differentiation.
Cancer immunology research (2015-11-26)
Kavitha Yaddanapudi, Beatriz E Rendon, Gwyneth Lamont, Eun Jung Kim, Numan Al Rayyan, Jamaal Richie, Sabrin Albeituni, Sabine Waigel, Ashley Wise, Robert A Mitchell
RESUMEN
Highly aggressive cancers "entrain" innate and adaptive immune cells to suppress antitumor lymphocyte responses. Circulating myeloid-derived suppressor cells (MDSC) constitute the bulk of monocytic immunosuppressive activity in late-stage melanoma patients. Previous studies revealed that monocyte-derived macrophage migration inhibitory factor (MIF) is necessary for the immunosuppressive function of tumor-associated macrophages and MDSCs in mouse models of melanoma. In the current study, we sought to determine whether MIF contributes to human melanoma MDSC induction and T-cell immunosuppression using melanoma patient-derived MDSCs and an ex vivo coculture model of human melanoma-induced MDSC. We now report that circulating MDSCs isolated from late-stage melanoma patients are reliant upon MIF for suppression of antigen-independent T-cell activation and that MIF is necessary for maximal reactive oxygen species generation in these cells. Moreover, inhibition of MIF results in a functional reversion from immunosuppressive MDSC to an immunostimulatory dendritic cell (DC)-like phenotype that is at least partly due to reductions in MDSC prostaglandin E(2) (PGE(2)). These findings indicate that monocyte-derived MIF is centrally involved in human monocytic MDSC induction/immunosuppressive function and that therapeutic targeting of MIF may provide a novel means of inducing antitumor DC responses in late-stage melanoma patients.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4-IPP, ≥97% (HPLC)