Merck
  • PCSK4-null sperm display enhanced protein tyrosine phosphorylation and ADAM2 proteolytic processing during in vitro capacitation.
PCSK4-null sperm display enhanced protein tyrosine phosphorylation and ADAM2 proteolytic processing during in vitro capacitation.
Fertility and sterility (2009-04-04)
Charles Gyamera-Acheampong, Julian Vasilescu, Daniel Figeys, Majambu Mbikay
RESUMEN
To study the molecular basis for the accelerated capacitation rate in PCSK4-null sperm. Comparative and controlled experimental research study. Academic medical institute. Male mice C57BL/6J wild-type or null congenics for the Pcsk4 allele. Cauda and epididymal sperm were capacitated for varying times. Differences in sperm protein tyrosine phosphorylation and proteolytic processing of sperm-egg ligands ADAM2 and ADAM3. The PCSK4-null sperm proteins are hyper-tyrosine phosphorylated during capacitation. This hyperphosphorylation is dependent on protein kinase A (PKA), albumin, and calcium. There is also more ADAM2 proteolytic processing from a 46-kDa form of ADAM2 to a 27-kDa form in PCSK4-null sperm than in wild-type sperm. This processing is dependent on cholesterol efflux. Lack of PCSK4 is associated with quantitative changes in the phosphorylation and proteolysis of sperm proteins during capacitation; therefore, alterations in signal transduction and proteolytic processing during capacitation may underlie the fertilization incompetence of PCSK4-null sperm. More investigation is needed to determine how and to what extent these changes might contribute to the loss of fertilizing ability of PCSK4-null sperm.