Merck
Effects of the Programmed Nutrition Beef Program on feedlot performance and carcass characteristics.
Journal of animal science (2015-05-29)
K J Phelps, J S Drouillard, J S Jennings, B E Depenbusch, C L Van Bibber-Krueger, K A Miller, M A Vaughn, D D Burnett, J M Gonzalez
RESUMEN
The objective of this study was to examine the effects of alternative finishing strategies on feedlot performance and carcass characteristics. Beef steers (64 pens; 8 steers/pen) were allocated to a randomized complete block design with a 2 × 2 factorial treatment arrangement. Factor 1 consisted of diet, with cattle fed a conventional (CON) diet or a diet consisting of Programmed Nutrition Beef Program (PN) supplements. The PN treatment included Programmed Nutrition Beef Receiver fed from d 1 through 20 of feeding and Programmed Nutrition Beef Finisher fed from d 21 to harvest. Factor 2 evaluated the presence (EGP+) or absence (EGP-) of exogenous growth promotants (ExGP) in the production system. Steers in the EGP+ treatments were initially implanted with Component E-S, reimplanted with Component TE-IS, and fed 400 mg·animal·dof ractopamine hydrochloride for the final 28 d before harvest. Steers were harvested on d 175, and strip loins were removed from 2 carcasses selected at random from each pen for transport to Kansas State University. One 1.27-cm-thick steak was removed from the anterior face for proximate and long-chain fatty acid analysis. There were no diet × ExGP interactions ( > 0.10) for feedlot performance except for DMI ( = 0.02). Steers in the PN/EGP+ treatment consumed more feed than all other treatments ( < 0.05). Both diet and ExGP affected DMI ( < 0.05), with PN and EGP+ steers consuming more feed than their contemporaries. Gain:feed and ADG were unaffected ( > 0.10) by diet, but ExGP improved these measures ( < 0.01). There were no diet × ExGP interactions for carcass characteristics except KPH fat and percentages of yield grade 3 and 4 carcasses ( < 0.05). Diet affected total incidence of liver abscesses because PN steers had a greater ( = 0.05) incidence of liver abscesses than steers in the CON treatment. Diet did not affect the other carcass characteristics ( > 0.10). Use of ExGP increased ( < 0.05) HCW, LM area, and 12th-rib fat but did not affect ( > 0.10) marbling score. Using ExGP reduced the percentage carcasses grading Premium Choice ( < 0.05). No diet × ExGP interactions or diet effects were detected for long-chain fatty acid profiles ( > 0.10). Use of ExGP increased ( < 0.05) the ratio of saturated:unsaturated fatty acids. In summary, the alternative feeding strategy presented in this study produced similar feedlot performance and carcass characteristics compared with a conventional feedlot system.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Methyl tridecanoate, ≥97% (GC)