Merck
Expression of transferrin receptor on murine hematopoietic progenitors.
Cellular immunology (1984-01-01)
J Lesley, R Hyman, R Schulte, J Trotter
RESUMEN
We have used a monoclonal antibody against the murine transferrin receptor to study the expression of the transferrin receptor on the hematopoietic progenitor cells (BFU-E, CFU-E, and CFU-C) present in mouse bone marrow. Elutriation and cell-sorting data are consistent with the hypothesis that most CFU-E are transferrin receptor positive while most BFU-E express much less transferrin receptor. CFU-C comprise both transferrin-receptor-positive and -negative cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
CD71 (MRQ-48) Mouse Monoclonal Antibody
Sigma-Aldrich
CD71 (EP232) Rabbit Monoclonal Primary Antibody