Merck
  • Quantum yields lower than unity in photo- induced dissociative electron transfers: the reductive cleavage of carbon tetrachloride.
Quantum yields lower than unity in photo- induced dissociative electron transfers: the reductive cleavage of carbon tetrachloride.
Chemphyschem : a European journal of chemical physics and physical chemistry (2000-12-15)
L Pause, M Robert, J M Savéant
RESUMEN
It has been shown recently that the electrochemical reduction of carbon tetrachloride in N,N'-dimethylformamide follows a mechanism in which electron transfer and bond cleavage are concerted. We report here results concerning photoinduced electron transfer from the singlet excited state of two aromatic molecules, 2-ethyl-9,10-dimethoxyanthracene and perylene, to CCl4 , which is characterised by a quantum yield of complete quenching fragmentation ranging from 0.7 to 0.8. It is shown that a quantum yield below unity is compatible with a dissociative mechanism and arises from partitioning of the system at the intersection of the product- and ground-state potential energy surfaces. This phenomenon predominates over back electron transfer from the clustered fragments state. The photoinduced reductive cleavage of CCl4 thus provides a clear illustration of the recent theoretical prediction, that photoinduced dissociative electron transfers are not necessarily endowed with a unity quantum yield. This offers an opportunity to estimate the magnitude of the electronic matrix element that couples the fragmented product state and the ground reactant state.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2-Ethyl-9,10-dimethoxyanthracene, 97%