Merck
  • Evidence for the involvement of vanilloid receptor in the antinociception produced by the dialdeydes unsaturated sesquiterpenes polygodial and drimanial in rats.
Evidence for the involvement of vanilloid receptor in the antinociception produced by the dialdeydes unsaturated sesquiterpenes polygodial and drimanial in rats.
Neuropharmacology (2004-02-21)
Eunice Andre, Juliano Ferreira, Angela Malheiros, Rosendo A Yunes, João B Calixto
RESUMEN
This study investigated whether or not the neonatal treatment of rats with the sesquiterpenes polygodial or drimanial could cause persistent antinociception similar to that induced by capsaicin. Rats were injected subcutaneously 48 h after birth with capsaicin (50 mg/kg), polygodial (150 mg/kg), drimanial (150 mg/kg) or vehicle (1ml/kg). Six to eight weeks later, rats were tested in models of nociception. Treatment of rats with capsaicin, polygodial or drimanial produced significant inhibition of the first phase and, to a lesser extent, the second phase of formalin-induced nociception. A significant reduction in Complete Freund's Adjuvant and capsaicin-induced hyperalgesia was observed in the animals neonatally treated with capsaicin, polygodial or drimanial compared with vehicle-treated rats. Moreover, both sesquiterpenes caused inhibition of plasma extravasation induced by injection of capsaicin. The neonatal treatment with capsaicin, polygodial or drimanial significantly decreased [3H]-resiniferatoxin binding sites in the rat spinal cord, but only capsaicin neonatal treatment significantly reduced the expression of TRPV1 in dorsal root ganglia (DRG) when assessed by Western blot. These results extend our previous findings demonstrating that the neonatal treatment of rats with polygodial or drimanial, similar to that reported for capsaicin, produced persistent antinociception in adult animals associated with TRPV1 down-regulation in the spinal cord, but not TRPV1 expression in DRG.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Polygodial, ≥97% (HPLC)