Merck
Glypican-3 expression and its relationship with recurrence of HCC after liver transplantation.
World journal of gastroenterology (2012-06-02)
Yu-Liang Wang, Zhi-Jun Zhu, Da-Hong Teng, Zhi Yao, Wei Gao, Zhong-Yang Shen
RESUMEN
To investigate the diagnostic value of glypican-3 (GPC3) and its relationship with hepatocellular carcinoma (HCC) recurrence after liver transplantation. HCC tissue samples (n = 31) obtained from patients who had undergone liver transplantation were analyzed. GPC3 mRNA and protein expression were analyzed by TaqMan real-time reverse transcription-polymerase chain reaction and immunohistochemistry. Correlation between the GPC3 expression and clinicopathological features was analyzed. The potential prognostic value of GPC3 was investigated by comparing recurrence-free survival between HCC patients with and without GPC3 expression. Using a cutoff value of 3.5 × 10⁻², 20 of 31 cancerous tissues had expression values of > 3.5 × 10⁻², whereas 3 of 31 adjacent non-neoplastic parenchyma and 0 of 20 control liver tissues had expression values of > 3.5 × 10⁻² (P < 0.001). GPC3 protein was immunoexpressed in 68% of cancerous tissues, but not in adjacent non-neoplastic parenchyma and control liver tissues. Vascular invasion was significantly related to GPC3 expression (P < 0.05). Recurrence-free survival was significantly longer for patients without GPC3 mRNA overexpression (> 3.5 × 10⁻²) and those without vascular invasion (P < 0.05 for both). GPC3 expression may serve as a valuable diagnostic marker for HCC. GPC3 mRNA overexpression may be an adverse indicator for HCC patients after liver transplantation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Glypican 3, recombinant, expressed in NSO cells