Merck
Effect of butyrate analogues on proliferation and differentiation in human neuroblastoma cell lines.
Anticancer research (1998-06-06)
P Rocchi, A M Ferreri, E Magrini, P Perocco
RESUMEN
Butyric acid has been shown in vitro to produce cytodifferentiation of a wide variety of neoplastic cells. The potential clinical use of this compound as a therapeutic agent is limited by its rapid metabolism. This has led to the examination, as potential antineoplastic agents, of compounds structurally correlated to butyrate, with longer biological half lives. In this study we investigated the effect in vitro of two butyrate analogues, tributyrin and butyramide, on inducing growth inhibition and expression of morphological and immunophenotypic properties, in human neuroblastoma cell lines. Treatment with tributyrin resulted in a strong inhibition of cell proliferation and in induction of extensive differentiation; on the contrary butyramide was scarcely effective or quite ineffective. These results demonstrate that tributyrin retains the effectiveness of butyrate and suggest that this analogue could have utility for cytodifferentiation therapy.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Butyramide, ≥98.0% (T)