Merck
Metabolism of 5-aminoquinoline with liver cytosol enzyme.
Indian journal of biochemistry & biophysics (1993-08-01)
R Banoo, D W Matheison
RESUMEN
In vitro oxidation and acetylation of 5-aminoquinoline by rabbit liver enzyme preparation has been investigated. Incubation of 5-aminoquinoline with cytosol fraction of the enzyme preparation in the presence of acetyl coenzyme-A gave rise to three different products, viz. 5-amino-2-hydroxy quinoline, 5-acetyl-aminoquinoline and 5-acetylamino-2-hydroxy quinoline due to the combined action of aldehyde oxidase and N-acetyl transferase. The metabolites thus obtained by enzyme catalysed reactions were isolated and separated by TLC, HPLC and subsequently characterised by IR and mass spectral fragmentation techniques.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
5-Aminoquinoline, 97%