Merck
Nijmegen-Bethesda assay to measure factor VIII inhibitors.
Methods in molecular biology (Clifton, N.J.) (2013-04-03)
Elizabeth Duncan, Margaret Collecutt, Alison Street
RESUMEN
Hemophilia A is an inherited bleeding disorder caused by a deficiency of factor VIII coagulant activity (FVIII:C). Patients are treated with infusions of either plasma-derived or recombinant factor VIII. However, some patients develop inhibitory antibodies (inhibitors) to infused factor VIII which render it ineffective. The original Bethesda method was developed to standardize measurement of inhibitors in a factor VIII neutralization assay. One Bethesda unit is defined as that amount of inhibitor that results in 50% residual FVIII:C activity of a defined test mixture. In the Nijmegen modification of the original Bethesda method, the pH and the protein concentration of the test mixture is further standardized. As a result, the FVIII:C in the test mixture is less prone to artifactual deterioration and the test has improved specificity. Even with a standardized procedure a number of factors can affect the performance of the test and it is important for laboratory staff to be aware of their impact on the result outcome.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Human coagulation factor VIII concentrate, European Pharmacopoeia (EP) Reference Standard