Merck
Chronic toxicity of styrene maleic anhydride, a male contraceptive, in rhesus monkeys (Macaca mulatta).
Contraception (1990-09-01)
N Sethi, R K Srivastava, R K Singh, G S Bhatia, N Sinha
RESUMEN
A newly developed male contraceptive, styrene maleic anhydride (SMA), was injected in the vas deferens of male rhesus monkeys for safety evaluation at the dose of 100 mg (contraceptive dose, CD), 250 mg (CD x 2.5) and 500 mg (CD x 5.0), and the monkeys were kept under observation for one year. The observed behavioural, haematological, biochemical and histopathological parameters in treated monkeys were comparable to controls. The results suggest the polymer SMA to be safe up to 5 times CD in monkeys.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Poly(styrene-alt-maleic acid) sodium salt solution, 13 wt. % in H2O