Merck
  • Pharmacokinetic study of methotrexate, folinic acid and their serum metabolites in children treated with high-dose methotrexate and leucovorin rescue.
Pharmacokinetic study of methotrexate, folinic acid and their serum metabolites in children treated with high-dose methotrexate and leucovorin rescue.
European journal of clinical pharmacology (1990-01-01)
C Wolfrom, R Hepp, R Hartmann, H Breithaupt, G Henze
RESUMEN
The pharmacokinetics of methotrexate (MTX), 7-hydroxymethotrexate (7-OHMTX), 2,4-diaminomethylpteroic acid (APA), folinic acid, and 5-methyltetrahydrofolate (5-MTHF) have been studied during 21 high-dose MTX (HDMTX) infusions (5 g.m-2 in 24 h) with leucovorin (LCV) rescue, a component of the therapy of 5 children with acute lymphoblastic leukemia (ALL). The median steady-state concentration of MTX was 66 mumol.l-1. Three elimination half-lifes were determined for MTX: 1.8 h, 6.4 h and a terminal 15 h. The median systemic MTX clearance was 110 mg.m-2.min-1. The 7-OHMTX level increased during each infusion and a Cmax of 19 mumol.l-1 was achieved at the end. Its initial half-life was 5 h and the terminal half-life was 12 h. Thus, the peak serum concentration ratio of 7-OHMTX to MTX was reached 24 h after the end of the infusion at a median ratio of 8. The MTX metabolite APA was detected in concentrations less than 0.06 mumol.l-1. The median folinic acid level during rescue, 48 h after starting the infusion, was 7.0 mumol.l-1 and 18 h following the last dose of LCV it was 0.44 mumol.l-1, leading to ratios of folinic acid to MTX of 31 and 6, respectively. The median 5-MTHF level during rescue was 0.44 mumol.l-1 with a median ratio of 5-MTHF to MTX of 2. Twenty infusions with 48 h MTX levels of less than 0.5 mumol.l-1 were without marked toxicity. Only one patient with a 48 h MTX concentration of 5.5 mumol.l-1 and a ratio of 5-MTHF to MTX of 0.08 suffered from ulcerating mucositis and septicaemia despite increased and prolonged LCV rescue.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Methotrexate impurity E, European Pharmacopoeia (EP) Reference Standard
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad