- Fluorophore-encapsulated solid-supported bilayer vesicles: a method for studying membrane permeation processes.
Fluorophore-encapsulated solid-supported bilayer vesicles: a method for studying membrane permeation processes.
Langmuir : the ACS journal of surfaces and colloids (2006-07-13)
Thomas L Williams, Margarida M L M Vareiro, A Toby A Jenkins
PMID16830985
RESUMEN
This letter describes a new method for studying the interaction of the membrane-lysing enzyme phospholipase A(2) (PLA(2)) with phospholipid bilayers by simultaneous measurements of enzyme binding and vesicle lysis using surface plasmon resonance (SPR) and permeabilization using surface plasmon field-enhanced fluorescence spectroscopy (SPFS). The PLA(2) inhibitor dimethyl-eicosadienoic acid was incorporated into the surface-bound vesicles and support bilayer in order to study its role in preventing PLA(2)-mediated vesicle lysis. This methodology has a generic applicability for the study of a range of membrane-disrupting agents.