- Enhanced two-photon excited fluorescence in three-dimensionally crosslinked bovine serum albumin microstructures.
Enhanced two-photon excited fluorescence in three-dimensionally crosslinked bovine serum albumin microstructures.
Optics express (2011-07-01)
Keng-Chi Cho, Chi-Hsiang Lien, Chun-Yu Lin, Chia-Yuan Chang, Lynn L H Huang, Paul J Campagnola, Chen Yuan Dong, Shean-Jen Chen
PMID21716404
RESUMEN
In this study, the intensity of two-photon excited fluorescence (TPEF) of xanthene dye, Rose Bengal (RB), was significantly enhanced via bovine serum albumin (BSA) microstructures fabricated by the two-photon crosslinking (TPC) technique. The RB was utilized as the photoactivator in the TPC processing and the enhanced TPEF intensity correlates with the concentration of fabricated crosslinked BSA microstructures via the power control and pulse selection of the employed femtosecond laser. As a result, fabrication of three-dimensional BSA microstructures can be simultaneously monitored by the use of TPEF intensity. The crosslinked BSA microstructures synthesized may be used as an ordered biomaterial for fluorescence enhancement.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Trimethylolpropane triacrylate, contains monomethyl ether hydroquinone as inhibitor, technical grade