Merck
Control of phlebotomine sandflies in confined spaces using diffusible repellents and insecticides.
Medical and veterinary entomology (2009-01-06)
M Sirak-Wizeman, R Faiman, A Al-Jawabreh, A Warburg
RESUMEN
The control of phlebotomine sandflies (Diptera: Psychodidae), the vectors of leishmaniasis, is directed mostly against adults as larvae develop in unknown or inaccessible habitats. In the current study we tested geraniol, a natural plant-derived product, as a space repellent and the synthetic pyrethroid prallethrin as a diffusible insecticide. Geraniol was dispersed in the air using diffusers with an electric fan and prallethrin was evaporated using electrically heated evaporators. Both substances were tested in inhabited bedrooms and in tents. Geraniol failed to effect significant reductions in the numbers of either Phlebotomus papatasi Scopoli in rooms or Phlebotomus sergenti Parrot in tents. In laboratory experiments, geraniol proved ineffective in preventing sandflies from feeding. By contrast, prallethrin was highly effective in reducing the number of sandflies in rooms as well as in tents. Exposure of sandflies to prallethrin in laboratory experiments caused 97% mortality rates. Both prallethrin and, to a lesser extent, geraniol reduced the number of Culex mosquitoes captured in tents. Electric liquid-vaporizers with 1.5% prallethrin are highly effective in protecting people from sandfly bites in confined spaces and may be useful in combating cutaneous leishmaniasis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Prallethrin, PESTANAL®, analytical standard