Merck
Identification of Candida albicans and C. tropicalis with an umbelliferyl-labelled galactosaminide.
Journal of medical microbiology (1995-09-01)
M F Simpanya
RESUMEN
The 4-methylumbelliferyl-N-acetyl-beta-D-galactosaminide (UAG) test was evaluated in parallel with chlamydospore (CHL) and germ-tube (GT) tests for the identification of Candida spp. The UAG test gave 86.6% correct identification of C. albicans and C. tropicalis. Non-C. albicans and non-C. tropicalis yeasts gave correct negative results in UAG tests. Only one isolate of Trichosporon beigelii gave a "false" positive reaction. The UAG test, which can be completed within 30 min, is a reliable test for screening non-C. albicans and non-C. tropicalis yeasts.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4-Methylumbelliferyl N-acetyl-β-D-galactosaminide, ≥98% (HPLC)