Merck
Enhanced host-guest electrochemical recognition of herbicide MCPA using a β-cyclodextrin carbon nanotube sensor.
Talanta (2012-09-13)
V Rahemi, J J Vandamme, J M P J Garrido, F Borges, C M A Brett, E M P J Garrido
RESUMEN
An electrochemical sensor for the determination of the chlorophenoxy herbicide MCPA has been developed, based on a combination of multi-walled carbon nanotubes with incorporated β-cyclodextrin and a polyaniline film modified glassy carbon electrode. The proposed molecular host-guest recognition based sensor has a high electrochemical sensitivity for the determination of MCPA. The electrochemical behaviour of MCPA at the chemically modified electrode was investigated in detail by cyclic voltammetry. The results indicate that the β-CD/MWCNT modified glassy carbon electrode exhibits efficient electrocatalytic oxidation of MCPA with high sensitivity, stability and lifetime. The analytical characteristics of this film were used for the quantitative determination of MCPA in natural waters. Cyclic voltammetry in phosphate buffer solution at pH 6.0, allowed the development of a method to determine MCPA, without any previous steps of extraction, clean-up, or derivatization, in the range of 10-100 μmol L(-1), with a detection limit of 0.99 μmol L(-1) in water. The results were statistically compared with those obtained through an established high-performance liquid chromatography technique, no significant differences having been found between the two methods.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4-Chloro-2-methylphenoxyacetic acid, technical, ≥95.0% (T)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
4-Chloro-2-methylphenoxyacetic acid, PESTANAL®, analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad