Merck
  • Eicosapentaenoic acid ethyl ester (AMR101) therapy in patients with very high triglyceride levels (from the Multi-center, plAcebo-controlled, Randomized, double-blINd, 12-week study with an open-label Extension [MARINE] trial).
Eicosapentaenoic acid ethyl ester (AMR101) therapy in patients with very high triglyceride levels (from the Multi-center, plAcebo-controlled, Randomized, double-blINd, 12-week study with an open-label Extension [MARINE] trial).
The American journal of cardiology (2011-06-21)
Harold E Bays, Christie M Ballantyne, John J Kastelein, Jonathan L Isaacsohn, Rene A Braeckman, Paresh N Soni
RESUMEN
AMR101 is an omega-3 fatty acid agent containing ≥96% eicosapentaenoic acid ethyl ester and no docosahexaenoic acid. Previous smaller studies suggested that highly purified eicosapentaenoic acid lowered triglyceride (TG) levels without increasing low-density lipoprotein (LDL) cholesterol levels. TG-lowering therapies such as fibrates, and fish oils containing both eicosapentaenoic acid and docosahexaenoic acid, can substantially increase LDL cholesterol levels when administered to patients with very high TG levels (≥500 mg/dl). The present double-blind study randomized 229 diet-stable patients with fasting TG ≥500 mg/dl and ≤2,000 mg/dl (with or without background statin therapy) to AMR101 4 g/day, AMR101 2 g/day, or placebo. The primary end point was the placebo-corrected median percentage of change in TG from baseline to week 12. The baseline TG level was 680, 657, and 703 mg/dl for AMR101 4 g/day, AMR101 2 g/day, and placebo. AMR101 4 g/day reduced the placebo-corrected TG levels by 33.1% (n = 76, p <0.0001) and AMR101 2 g/day by 19.7% (n = 73, p = 0.0051). For a baseline TG level >750 mg/dl, AMR101 4 g/day reduced the placebo-corrected TG levels by 45.4% (n = 28, p = 0.0001) and AMR101 2 g/day by 32.9% (n = 28, p = 0.0016). AMR101 did not significantly increase the placebo-corrected median LDL cholesterol levels at 4 g/day (-2.3%) or 2 g/day (+5.2%; both p = NS). AMR101 significantly reduced non-high-density lipoprotein cholesterol, apolipoprotein B, lipoprotein-associated phospholipase A(2), very low-density lipoprotein cholesterol, and total cholesterol. AMR101 was generally well tolerated, with a safety profile similar to that of the placebo. In conclusion, the present randomized, double-blind trial of patients with very high TG levels demonstrated that AMR101 significantly reduced the TG levels and improved other lipid parameters without significantly increasing the LDL cholesterol levels.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Eicosapentaenoic acid ethyl ester, European Pharmacopoeia (EP) Reference Standard