Merck
  • New approach to large-volume injection in reversed-phase high performance liquid chromatography: determination of atrazine and hydroxyatrazine in soil samples.
New approach to large-volume injection in reversed-phase high performance liquid chromatography: determination of atrazine and hydroxyatrazine in soil samples.
Journal of separation science (2006-10-05)
Milan Hutta, Mária Chalányová, Radoslav Halko, Róbert Góra, Ivan Rybár, Marek Pajchl, Svetlana Dokupilová
RESUMEN
A well established method of direct injection of larger than conventional sample volumes ranging from 0.1 mL to 10 mL in HPLC is the injection valve method in which a loop of tubing is totally or partially filled with sample. Recent HPLC pumps have a flow-rate setting accuracy of +/- 1-2% over a flow-rate range from 0.1 mL/min to 10 mL/min and the flow stability is 0.2% or less. Quarternary low pressure gradient pumps are widely available and used, but all their hydraulic lines are seldom utilised. The idea of using one line of a common commercial HPLC quaternary low-pressure pump for direct on-column injection (pumping) of large sample volumes ranging from 1 mL to 100 mL was tested. This approach was evaluated during practical work on the development of an RP-HPLC method of determination of residual atrazine and hydroxyatrazine. In lysimetric environmental experiments hydroxyatrazine was formed in situ in a soil column by hydrolysis of atrazine. The results proved the applicability of this approach not only in experiments with model mixtures of analytes at microg/L levels in solutions. Analysis of 20 mL of soil leachates and extracts of soil samples containing atrazine and hydroxyatrazine at the 10 microg/kg level (in dry soil) revealed that good figures-of-merit were preserved, even in the presence of a large excess of humic substances.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Atrazine-2-hydroxy, PESTANAL®, analytical standard