Merck
  • Microbial transformation of 17alpha-ethynyl- and 17alpha-ethylsteroids, and tyrosinase inhibitory activity of transformed products.
Microbial transformation of 17alpha-ethynyl- and 17alpha-ethylsteroids, and tyrosinase inhibitory activity of transformed products.
Steroids (2005-08-02)
M Iqbal Choudhary, Sadia Sultan, Mahmud Tareq Hassan Khan, Atta-Ur- Rahman
RESUMEN
The microbial transformation of the 17alpha-ethynyl-17beta-hydroxyandrost-4-en-3-one (1) (ethisterone) and 17alpha-ethyl-17beta-hydroxyandrost-4-en-3-one (2) by the fungi Cephalosporium aphidicola and Cunninghamella elegans were investigated. Incubation of compound 1 with C. aphidicola afforded oxidized derivative, 17alpha-ethynyl-17beta-hydroxyandrosta-1,4-dien-3-one (3), while with C. elegans afforded a new hydroxy derivative, 17alpha-ethynyl-11alpha,17beta-dihydroxyandrost-4-en-3-one (4). On the other hand, the incubation of compound 2 with the fungus C. aphidicola afforded 17alpha-ethyl-17beta-hydroxyandrosta-1,4-dien-3-one (5). Two new hydroxylated derivatives, 17alpha-ethyl-11alpha,17beta-dihydroxyandrost-4-en-3-one (6) and 17alpha-ethyl-6alpha,17beta-dihydroxy-5alpha-androstan-3-one (7) were obtained from the incubation of compound 2 with C. elegans. Compounds 1-6 exhibited tyrosinase inhibitory activity, with compound 6 being the most potent member (IC(50)=1.72 microM).
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ethisterone, ≥99% (HPLC)