Merck
  • Acyl-CoA synthetase 6 controls rod photoreceptor function and survival by shaping the phospholipid composition of retinal membranes.
Acyl-CoA synthetase 6 controls rod photoreceptor function and survival by shaping the phospholipid composition of retinal membranes.
Communications biology (2024-08-22)
Yixiao Wang, Silke Becker, Stella Finkelstein, Frank M Dyka, Haitao Liu, Mark Eminhizer, Ying Hao, Richard S Brush, William J Spencer, Vadim Y Arshavsky, John D Ash, Jianhai Du, Martin-Paul Agbaga, Frans Vinberg, Jessica M Ellis, Ekaterina S Lobanova
RESUMEN
The retina is light-sensitive neuronal tissue in the back of the eye. The phospholipid composition of the retina is unique and highly enriched in polyunsaturated fatty acids, including docosahexaenoic fatty acid (DHA). While it is generally accepted that a high DHA content is important for vision, surprisingly little is known about the mechanisms of DHA enrichment in the retina. Furthermore, the biological processes controlled by DHA in the eye remain poorly defined as well. Here, we combined genetic manipulations with lipidomic analysis in mice to demonstrate that acyl-CoA synthetase 6 (Acsl6) serves as a regulator of the unique composition of retinal membranes. Inactivation of Acsl6 reduced the levels of DHA-containing phospholipids, led to progressive loss of light-sensitive rod photoreceptor neurons, attenuated the light responses of these cells, and evoked distinct transcriptional response in the retina involving the Srebf1/2 (sterol regulatory element binding transcription factors 1/2) pathway. This study identifies one of the major enzymes responsible for DHA enrichment in the retinal membranes and introduces a model allowing an evaluation of rod functioning and pathology caused by impaired DHA incorporation/retention in the retina.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-ACSL6 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution