Merck
Structure features of peptide-type SARS-CoV main protease inhibitors: Quantitative structure activity relationship study.
Chemometrics and intelligent laboratory systems : an international journal sponsored by the Chemometrics Society (2021-02-02)
Vijay H Masand, Siddhartha Akasapu, Ajaykumar Gandhi, Vesna Rastija, Meghshyam K Patil
RESUMEN
In the present work, an extensive QSAR (Quantitative Structure Activity Relationships) analysis of a series of peptide-type SARS-CoV main protease (MPro) inhibitors following the OECD guidelines has been accomplished. The analysis was aimed to identify salient and concealed structural features that govern the MPro inhibitory activity of peptide-type compounds. The QSAR analysis is based on a dataset of sixty-two peptide-type compounds which resulted in the generation of statistically robust and highly predictive multiple models. All the developed models were validated extensively and satisfy the threshold values for many statistical parameters (for e.g. R2 ​= ​0.80-0.82, Q2 loo ​= ​0.74-0.77, Q 2 LMO  ​= ​0.66-0.67). The developed QSAR models identified number of sp2 hybridized Oxygen atoms within seven bonds from aromatic Carbon atoms, the presence of Carbon and Nitrogen atoms at a topological distance of 3 and other interrelations of atom pairs as important pharmacophoric features. Hence, the present QSAR models have a good balance of Qualitative (Descriptive QSARs) and Quantitative (Predictive QSARs) approaches, therefore useful for future modifications of peptide-type compounds for anti- SARS-CoV activity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Mpro inhibitor 5h, ≥98% (HPLC)